Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human FABP2/I-FABP Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF3078
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF3078 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF3078 Supplier R&D Systems Supplier No. BAF3078
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

FABP2/I-FABP Polyclonal specifically detects FABP2/I-FABP in Human samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).

Specifications

Antigen FABP2/I-FABP
Applications Western Blot, ELISA
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL
Gene Accession No. P12104
Gene Alias FABPI, fatty acid binding protein 2, intestinal, Fatty acid-binding protein 2, fatty acid-binding protein, intestinal, IFABP, I-FABP, I-FABPMGC133132, Intestinal FABP, Intestinal-type fatty acid-binding protein
Gene Symbols FABP2
Host Species Goat
Immunogen E. coli-derived recombinant human FABP2/I-FABP Ala2-Asp132 Accession # P12104
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2169
Test Specificity Detects human FABP2/I-FABP in ELISAs and Western blots. In sandwich immunoassays, less than 0.4% cross-reactivity with recombinant rat FABP2 is observed, and less than 0.1% cross-reactivity with recombinant human (rh) FABP1, -3, -5, -6, -7, -8, rhFABP9, recombinant mouse (rm) FABP4, and rmFABP9 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.