Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human FABP4/A-FABP Antibody, R&D Systems™
GSA_VAGREENER_CHOICE

Catalog No. AF3150
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF3150 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF3150 Supplier R&D Systems Supplier No. AF3150
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 8 publications

FABP4/A-FABP Polyclonal specifically detects FABP4/A-FABP in Human samples. It is validated for Western Blot, Simple Western, ELISA, Immunohistochemistry.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Antigen FABP4/A-FABP
Applications Western Blot, ELISA, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Simple Western 10 ug/mL, ELISA, Immunohistochemistry 5-15 ug/mL
Gene Accession No. P15090
Gene Alias Adipocyte lipid-binding protein, Adipocyte-type fatty acid-binding protein, AFABP, A-FABP, A-FABPAFABP, ALBP, aP2, fatty acid binding protein 4, adipocyte, Fatty acid-binding protein 4, fatty acid-binding protein, adipocyte
Gene Symbols FABP4
Host Species Goat
Immunogen E. coli-derived recombinant human FABP4/A-FABP Cys2-Ala132 Accession # P15090
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2167
Test Specificity Detects human FABP4/A-FABP in direct ELISAs and Western blots. In direct ELISAs, approximately 60% cross-reactivity with recombinant mouse (rm) FABP4 is observed, approximately 30% cross-reactivity with recombinant human (rh) FABP3 is observed, and less than 5% cross-reactivity with rhFABP1, -2, -5, -6, -7, -8, -9, and rmFABP9 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.