Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human FABP5/E-FABP Alexa Fluor 405-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems FAB3077V100UG
This item is not returnable.
View return policy
Description
FABP5/E-FABP Monoclonal specifically detects FABP5/E-FABP in Human samples. It is validated for Flow Cytometry.Specifications
FABP5/E-FABP | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q01469 | |
FABP5 | |
E. coli-derived recombinant human FABP5/E-FABP Ala2-Glu135 Accession # Q01469 | |
RUO | |
2171 | |
Human | |
IgG2a |
Flow Cytometry | |
311215 | |
Alexa Fluor 405 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
EFABP, E-FABP, E-FABPPAFABP, fatty acid binding protein 5 (psoriasis-associated), Fatty acid-binding protein 5, PA-FABPepidermal, Psoriasis-associated fatty acid-binding protein homolog | |
Rat | |
100 μg | |
Primary | |
Detects human FABP5/E-FABP in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human FABP1, 2, 3, 4, 6, 7, or 9 is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction