Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human FABP5/E-FABP Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF3077
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF3077 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF3077 Supplier R&D Systems Supplier No. BAF3077
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

FABP5/E-FABP Polyclonal specifically detects FABP5/E-FABP in Human samples. It is validated for Western Blot.

Specifications

Antigen FABP5/E-FABP
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL
Gene Accession No. Q01469
Gene Alias EFABP, E-FABP, E-FABPPAFABP, fatty acid binding protein 5 (psoriasis-associated), Fatty acid-binding protein 5, PA-FABPepidermal, Psoriasis-associated fatty acid-binding protein homolog
Gene Symbols FABP5
Host Species Goat
Immunogen E. coli-derived recombinant human FABP5/E-FABP Ala2-Glu135 Accession # Q01469
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2171
Test Specificity Detects human FABP5/E-FABP in Western blots. In Western blots, approximately 30% cross-reactivity with recombinant mouse (rm) FABP5 is observed and less than 5% cross-reactivity with recombinant human (rh) FABP1, rhFABP2, rhFABP3, rhFABP4, rhFABP6, rhFABP7, and rmFABP9 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.