Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human FABP6 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF3880
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF3880 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF3880 Supplier R&D Systems Supplier No. AF3880
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Produced in goats immunized with purified, E. coli-derived, recombinant human FABP6 (rhFABP6; aa 2 - 128; Accession # P51161). Human FABP6 specific IgG was purified by human FABP6 affinity chromatography.

FABP6 Polyclonal specifically detects FABP6 in Human samples. It is validated for Western Blot, Immunocytochemistry.

Specifications

Antigen FABP6
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL
Gene Accession No. P51161
Gene Alias fatty acid binding protein 6, ileal, Fatty acid-binding protein 6, gastrotropin, I-15P, I-15PIntestinal 15 kDa protein, IBABP, I-BABPIntestinal bile acid-binding protein, I-BALB, I-BAP, ILBP, ILBP3, ILBPGT, ileal bile acid binding protein, ILLBP, ILLBPIleal lipid-binding protein, illeal lipid-binding protein
Gene Symbols FABP6
Host Species Goat
Immunogen E. coli-derived recombinant human FABP6 Ala2-Ala128 Accession # P51161
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2172
Test Specificity Detects human FABP6 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant human (rh) FABP1, rhFABP2, rhFABP3, rhFABP4, rhFABP5, rhFABP7, and recombinant mouse FABP9 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.