Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human FABP8/M-FABP Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF5866SP
This item is not returnable.
View return policy
Description
FABP8/M-FABP/Myelin P2 Protein Polyclonal specifically detects FABP8/M-FABP/Myelin P2 Protein in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
FABP8/M-FABP/Myelin P2 Protein | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
MFABP, M-FABP, MP2, myelin P2 protein, P2, peripheral myelin protein 2FABP8, PMP2 | |
Sheep | |
Affinity Purified | |
RUO | |
5375 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
P02689 | |
PMP2 | |
E. coli-derived recombinant human FABP8 Ser2-Val132 Accession # P02689 | |
25 μg | |
Primary | |
Detects human FABP8 in direct ELISAs and Western blots. In direct ELISAs, less than 2% cross-reactivity with recombinant human (rh) FABP2, 3, 4, 5, 6, 7 and 9 is observed. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction