Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human FABP8/M-FABP Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB5866
This item is not returnable.
View return policy
Description
FABP8/M-FABP/Myelin P2 Protein Monoclonal specifically detects FABP8/M-FABP/Myelin P2 Protein in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
FABP8/M-FABP/Myelin P2 Protein | |
Monoclonal | |
LYOPH | |
Western Blot 2 ug/mL, Immunohistochemistry 8-25 ug/mL | |
P02689 | |
PMP2 | |
E. coli-derived recombinant human FABP8/M-FABP Ser2-Val132 Accession # P02689 | |
100 μg | |
Primary | |
Detects human FABP8 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human FABP1, 2, 3, 4, 5, 6, 7, or 9 is observed. | |
Human | |
Purified |
Western Blot, Immunohistochemistry | |
631306 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
MFABP, M-FABP, MP2, myelin P2 protein, P2, peripheral myelin protein 2FABP8, PMP2 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
5375 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction