Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human FABP8/M-FABP Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB5866SP
This item is not returnable.
View return policy
Description
FABP8/M-FABP/Myelin P2 Protein Monoclonal specifically detects FABP8/M-FABP/Myelin P2 Protein in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
FABP8/M-FABP/Myelin P2 Protein | |
Monoclonal | |
LYOPH | |
Western Blot 2 ug/mL, Immunohistochemistry 8-25 ug/mL | |
P02689 | |
PMP2 | |
E. coli-derived recombinant human FABP8/M-FABP Ser2-Val132 Accession # P02689 | |
25 μg | |
Primary | |
Detects human FABP8 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human FABP1, 2, 3, 4, 5, 6, 7, or 9 is observed. | |
Human | |
Purified |
Western Blot, Immunohistochemistry | |
631306 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
MFABP, M-FABP, MP2, myelin P2 protein, P2, peripheral myelin protein 2FABP8, PMP2 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
5375 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction