Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human FABP8/M-FABP Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF5866
This item is not returnable.
View return policy
Description
FABP8/M-FABP/Myelin P2 Protein Polyclonal specifically detects FABP8/M-FABP/Myelin P2 Protein in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
FABP8/M-FABP/Myelin P2 Protein | |
Polyclonal | |
Unconjugated | |
P02689 | |
PMP2 | |
E. coli-derived recombinant human FABP8 Ser2-Val132 Accession # P02689 | |
100 μg | |
Primary | |
Detects human FABP8 in direct ELISAs and Western blots. In direct ELISAs, less than 2% cross-reactivity with recombinant human (rh) FABP2, 3, 4, 5, 6, 7 and 9 is observed. | |
Human | |
IgG |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
MFABP, M-FABP, MP2, myelin P2 protein, P2, peripheral myelin protein 2FABP8, PMP2 | |
Sheep | |
Affinity Purified | |
RUO | |
5375 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction