Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human FABP8/M-FABP/Myelin P2 Protein ELISA Kit (Colorimetric)
SDP

Catalog No. NB013334 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB013334 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB013334 Supplier Novus Biologicals™ Supplier No. NBP280263
Add to Cart
Add to Cart

This kit recognizes Human P2 in samples.

No significant cross-reactivity or interference between Human P2 and analogues was observed.

Specifications

Accession Number P02689, P02689
Assay Range 15.625 to 1000 pg/mL
Assay Sensitivity 9.375 pg/mL
Product Type ELISA Kit (Colorimetric)
Sample Type Other Biological Fluids, Plasma, Serum
For Use With (Application) ELISA
Gene ID (Entrez) 5375
Gene Symbol PMP2
Host Species Human
Quantity 1 Kit
Storage Requirements Storage of components varies. See protocol for specific instructions.
Target FABP8/M-FABP/Myelin P2 Protein
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.