Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human FACA/FANCA Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF6026SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF6026SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF6026SP Supplier R&D Systems Supplier No. AF6026SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

FACA/FANCA Polyclonal specifically detects FACA/FANCA in Human samples. It is validated for Western Blot.

Specifications

Antigen FACA/FANCA
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. O15360
Gene Alias FA, FA1, FAA, FAAMGC75158, FACAFanconi anemia group A protein, FAH, FA-H, FANCA, FANCH, Fanconi anemia, complementation group A, Fanconi anemia, complementation group H, Fanconi anemia, type 1, Protein FACA
Gene Symbols FANCA
Host Species Goat
Immunogen E. coli-derived recombinant human FACA/FANCA Tyr35-Arg142 Accession # O15360
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2175
Test Specificity Detects human FACA/FANCA in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant mouse FANCA, recombinant human (rh) FANCD2, rhFANCE, rhFANCF, rhFANCG, and rhFANCI is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.