Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human FACA/FANCA Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF6026
This item is not returnable.
View return policy
Description
FACA/FANCA Polyclonal specifically detects FACA/FANCA in Human samples. It is validated for Western Blot.Specifications
FACA/FANCA | |
Polyclonal | |
Unconjugated | |
O15360 | |
FANCA | |
E. coli-derived recombinant human FACA/FANCA Tyr35-Arg142 Accession # O15360 | |
100 μg | |
Primary | |
Detects human FACA/FANCA in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant mouse FANCA, recombinant human (rh) FANCD2, rhFANCE, rhFANCF, rhFANCG, and rhFANCI is observed. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
FA, FA1, FAA, FAAMGC75158, FACAFanconi anemia group A protein, FAH, FA-H, FANCA, FANCH, Fanconi anemia, complementation group A, Fanconi anemia, complementation group H, Fanconi anemia, type 1, Protein FACA | |
Goat | |
Affinity Purified | |
RUO | |
2175 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction