Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human Factor VIII ELISA Kit (Colorimetric)
SDP

Catalog No. NB170562 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB170562 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB170562 Supplier Novus Biologicals™ Supplier No. NBP318716
Add to Cart
Add to Cart
This item is not returnable. View return policy

Colorimetric ELISA Kit

Specifications

Assay Range 0.375 - 24 ng/mL; 2.5 - 160 mIU/mL (example only; lot dependent)
Assay Sensitivity 0.21 ng/mL (example only; lot dependent)
Product Type ELISA Kit (Colorimetric)
Sample Type Breast Milk, Cell Culture, Cerebrospinal Fluid, Plasma, Serum, Tissue Extracts
For Use With (Application) ELISA
Gene ID (Entrez) 2157
Gene Symbol F8
Quantity 1 Kit
Research Discipline Cardiovascular Biology
Storage Requirements Storage is content dependent.
Synonym AHF, Antihemophilic factor, coagulation factor VIII, procoagulant component, coagulation factor VIIIc, DXS1253E, F8Ccoagulation factor VIII, factor VIII F8B, FVIII, HEMAF8B, Procoagulant component
Target Human
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.