Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human FAM3A Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody

$188.28 - $473.38

Specifications

Antigen FAM3A
Clone 358112
Concentration LYOPH
Dilution Western Blot 1 ug/mL
Applications Western Blot
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB2865SP
GREENER_CHOICE
View Documents
R&D Systems
MAB2865SP
25 μg
Each for $188.28
Only null left
Add to Cart
 
MAB2865
GREENER_CHOICE
View Documents
R&D Systems
MAB2865
100 μg
Each for $473.38
Only null left
Add to Cart
 
Description

Description

FAM3A Monoclonal specifically detects FAM3A in Human samples. It is validated for Western Blot.
Specifications

Specifications

FAM3A
LYOPH
Western Blot
Unconjugated
Mouse
Human
P98173
60343
Mouse myeloma cell line NS0-derived recombinant human FAM3A Phe34-Ser230 Accession # P98173
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human FAM3A in direct ELISAs and Western blots. In direct ELISAs and Western blots, 100% cross-reactivity with recombinant mouse FAM3A is observed and no cross-reactivity with recombinant human (rh) FAM3B, rhFAM3C, or rhFAM3D is observed.
358112
Western Blot 1 ug/mL
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
2.19, Cytokine-like protein 2-19, DLD, DXS560S, family with sequence similarity 3, member A, member A protein, XAP-7
FAM3A
IgG2b
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.