Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human FAM3B Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $468.00
Specifications
Antigen | FAM3B |
---|---|
Clone | 365505 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Description
FAM3B Monoclonal specifically detects FAM3B in Human samples. It is validated for Western Blot.Specifications
FAM3B | |
LYOPH | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
NP_478066 | |
54097 | |
Mouse myeloma cell line NS0-derived recombinant human FAM3B isoform B Glu30-Ser235 Accession # NP_478066 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human FAM3B in direct ELISAs and Western blots. In direct ELISAs and Western blots, 75-100% cross-reactivity with recombinant mouse FAM3B and no cross-reactivity with recombinant human (rh) FAM3A, rhFAM3C, or rhFAM3D is observed. |
365505 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
2-21, C21orf11, C21orf76PANDER, chromosome 21 open reading frame 11, Cytokine-like protein 2-21, D21M16SJHU19e, family with sequence similarity 3, member B, ORF9, pancreatic derived factor, Pancreatic-derived factor, PANDER, PRED44 | |
FAM3B | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title