Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human FAM3D Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF2869
This item is not returnable.
View return policy
Description
FAM3D Polyclonal specifically detects FAM3D in Human samples. It is validated for Western Blot.Specifications
FAM3D | |
Polyclonal | |
Unconjugated | |
Q96BQ1 | |
FAM3D | |
Mouse myeloma cell line NS0-derived recombinant human FAM3D Trp39-Phe224 Accession # Q96BQ1 | |
100 μg | |
Primary | |
Detects human FAM3D in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant mouse FAM3D is observed and 5% cross-reactivity with recombinant human (rh) FAM3A, rhFAM3B, and rhFAM3C is observed. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
cytokine-like protein EF-7, EF7, family with sequence similarity 3, member D, OIT1 | |
Goat | |
Affinity Purified | |
RUO | |
131177 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction