Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human FANCD2 Alexa Fluor 750-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Supplier: R&D Systems IC9369S100UG
This item is not returnable.
View return policy
Description
FANCD2 Monoclonal specifically detects FANCD2 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
FANCD2 | |
Monoclonal | |
Alexa Fluor 750 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
DKFZp762A223, FA4, FACD, FAD, FAD2, FA-D2, FADFAD2, FANCD, Fanconi anemia complementation group D2, Fanconi anemia group D2 protein, Fanconi anemia, complementation group D2, FLJ23826, FPN1, HFE4, IREG1, Protein FACD2, SLC11A3 | |
E. coli-derived recombinant human FANCD2, Glu11-Glu230, Accession # Q9BXW9 | |
100 μg | |
Primary | |
Detects human FANCD2 in direct ELISAs and Western blots. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG |
Flow Cytometry | |
1290D | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
Q9BXW9 | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
RUO | |
2177 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction