Learn More
Description
The Quantikine QuicKit Human FAP Immunoassay is a one-step, quantitative colorimetric ELISA designed to measure human FAP in cell culture supernates, serum, and plasma.
- Assay Length: 80 minutes
- Assay Range: 54.7 to 3500 pg/mL
- Sensitivity: 9.01 pg/mL
- Detection Method: Colorimetric (TMB, 450 nm)
- Conjugate: HRP
- Specificity: Detects natural and recombinant human FAP
- Sample Requirements: 50 μL (Cell Culture Supernates), 13 μL (Serum and Plasma)
- Research Area: Cancer Biomarkers
Specifications
Specifications
| Product Type | ELISA Kit |
| Quantity | 1 Kit |
| Species | Human |
| Binding Target | Fibroblast Activation Protein alpha/FAP |
| Assay Sensitivity | 9.01 pg/mL |
| Assay Range | 54.70 - 3500 pg/mL |
| Gene ID (Entrez) | 2191 |
| Product Line | R&D Systems |
| Sample Type | Cell Culture Supernates (50 uL), Serum (13 uL), EDTA Plasma (13 uL), Heparin Plasma (13 uL) |
| Synonym | 170 kDa melanoma membrane-bound gelatinase, DKFZp686G13158, DPPIV, EC 3.4.21.-, FAP, FAPA, Fibroblast activation protein alpha, fibroblast activation protein, alpha, Integral membrane serine protease, seprase, vibronectin |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
