Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Fas/TNFRSF6/CD95 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB142
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB142 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB142 Supplier R&D Systems Supplier No. MAB142
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 4 publications

Fas/TNFRSF6/CD95 Monoclonal specifically detects Fas/TNFRSF6/CD95 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Fas/TNFRSF6/CD95
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone DX2
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Alias Apo-1, Apo-1 antigen, apoptosis antigen 1, Apoptosis-mediating surface antigen FAS, APT1, APT1FASTM, CD95, CD95 antigen, CD95ALPS1A, Fas (TNF receptor superfamily, member 6), Fas AMA, Fas antigen, FAS1, FASLG receptor, TNFRSF6, TNFRSF6member 6, tumor necrosis factor receptor superfamily member 6
Gene Symbols FAS
Host Species Mouse
Immunogen L cells transfected with human Fas
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 355
Test Specificity Detects human Fas in direct ELISAs and Western blots. Does not cross-react with rhCD27, rmCD27, rhCD30, rmCD30, rhCD40, rmCD40, rhDR3, rmDR3, rhHVEM, rmHVEM, rhLymphotoxin beta R, rmLymphotoxin beta R, rhTRAIL R1, rmTRAIL R1, rhTRAIL R2, rmTRAIL R2, rhTRAIL R3, or rhTRAIL R4,
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.