Learn More
Description
Specifications
Specifications
| Assay Range | 0.78 -50ng/mL |
| Assay Sensitivity | 0.32ng/mL |
| Product Type | ELISA Kit |
| Target Species | Human |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 6624 |
| Gene Symbol | FSCN1 |
| Quantity | 1 Kit |
| Research Discipline | Cytoskeleton Markers |
| Storage Requirements | Storage is content dependent. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
