Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human FATP2 Alexa Fluor 647-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. IC4659R100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC4659R100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC4659R100 Supplier R&D Systems Supplier No. IC4659R100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

FATP2/SLC27A2 Monoclonal specifically detects FATP2/SLC27A2 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen FATP2/SLC27A2
Applications Flow Cytometry
Classification Monoclonal
Clone 466106
Conjugate Alexa Fluor 647
Dilution Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Accession No. O14975
Gene Alias ACSVL1, ACSVL1FATP-2, EC 6.2.1, EC 6.2.1.-, EC 6.2.1.3, EC 6.2.1.7, FACVL1, FACVL1very long-chain fatty-acid-coenzyme A ligase 1, FATP2THCA-CoA ligase, Fatty acid transport protein 2, Fatty-acid-coenzyme A ligase, very long-chain 1, hFACVL1, HsT17226, Long-chain-fatty-acid--CoA ligase, SLC27A2, solute carrier family 27 (fatty acid transporter), member 2, Solute carrier family 27 member 2, THCA-CoA ligase, very-long-chain acyl-CoA synthetase, VLACS, VLACSVery long-chain-fatty-acid-CoA ligase, VLCS, VLCSvery long-chain acyl-CoA synthetase
Host Species Mouse
Immunogen HEK293 human embryonic kidney cell line transfected with human FATP2 Met1-Leu620 Accession # O14975
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11001
Test Specificity Detects human FATP2. Stains human FATP2 transfectants but not irrelevant transfectants.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.