Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human FATP2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB4659
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4659 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB4659 Supplier R&D Systems Supplier No. MAB4659
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

FATP2/SLC27A2 Monoclonal specifically detects FATP2/SLC27A2 in Human samples. It is validated for Intracellular Staining by Flow Cytometry, CyTOF-ready.

Specifications

Antigen FATP2/SLC27A2
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 466106
Concentration LYOPH
Conjugate Unconjugated
Dilution Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. O14975
Gene Alias ACSVL1, ACSVL1FATP-2, EC 6.2.1, EC 6.2.1.-, EC 6.2.1.3, EC 6.2.1.7, FACVL1, FACVL1very long-chain fatty-acid-coenzyme A ligase 1, FATP2THCA-CoA ligase, Fatty acid transport protein 2, Fatty-acid-coenzyme A ligase, very long-chain 1, hFACVL1, HsT17226, Long-chain-fatty-acid--CoA ligase, SLC27A2, solute carrier family 27 (fatty acid transporter), member 2, Solute carrier family 27 member 2, THCA-CoA ligase, very-long-chain acyl-CoA synthetase, VLACS, VLACSVery long-chain-fatty-acid-CoA ligase, VLCS, VLCSvery long-chain acyl-CoA synthetase
Gene Symbols SLC27A2
Host Species Mouse
Immunogen HEK293 human embryonic kidney cell line transfected with human FATP2 Met1-Leu620 Accession # O14975
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11001
Test Specificity Detects human FATP2. Stains human FATP2 transfectants but not irrelevant transfectants.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.