Learn More
Human FATP5 Alexa Fluor 405-conjugated Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | FATP5/SLC27A5 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 390917 |
| Concentration | 0.2 mg/mL |
| Conjugate | Alexa Fluor 405 |
| Dilution | Flow Cytometry 0.25-1 ug/10^6 cells |
| Formulation | Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide |
| Gene Accession No. | Q9Y2P5 |
| Gene Alias | ACSB, ACSBBACS, ACSVL6, ACSVL6BAL, BA-CoA ligase, Bile acid-CoA ligase, bile acyl-CoA synthetase, Cholate--CoA ligase, EC 6.2.1.7, FACVL3, FACVL3FATP-5, FATP5Solute carrier family 27 member 5, Fatty acid transport protein 5, Fatty-acid-coenzyme A ligase, very long-chain 3, FLJ22987, SLC27A5, solute carrier family 27 (fatty acid transporter), member 5, VLACSR, VLACSRVery long-chain acyl-CoA synthetase homolog 2, VLCSH2, VLCSH2Very long-chain acyl-CoA synthetase-related protein, VLCS-H2VLACS-related |
| Show More |
For Research Use Only (RUO)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.