Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human FATP5 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB3897
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3897 100 μg
MAB3897SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB3897 Supplier R&D Systems Supplier No. MAB3897
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

FATP5/SLC27A5 Monoclonal specifically detects FATP5/SLC27A5 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.

Specifications

Antigen FATP5/SLC27A5
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 390917
Concentration LYOPH
Conjugate Unconjugated
Dilution Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9Y2P5
Gene Alias ACSB, ACSBBACS, ACSVL6, ACSVL6BAL, BA-CoA ligase, Bile acid-CoA ligase, bile acyl-CoA synthetase, Cholate--CoA ligase, EC 6.2.1.7, FACVL3, FACVL3FATP-5, FATP5Solute carrier family 27 member 5, Fatty acid transport protein 5, Fatty-acid-coenzyme A ligase, very long-chain 3, FLJ22987, SLC27A5, solute carrier family 27 (fatty acid transporter), member 5, VLACSR, VLACSRVery long-chain acyl-CoA synthetase homolog 2, VLCSH2, VLCSH2Very long-chain acyl-CoA synthetase-related protein, VLCS-H2VLACS-related
Gene Symbols SLC27A5
Host Species Mouse
Immunogen HEK293 human embryonic kidney cell line transfected with human FATP5 Met1-Leu690 Accession # Q9Y2P5
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10998
Test Specificity Detects human FATP5. Stains human FATP5 transfectants but not irrelevant transfectants.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.