Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Fatty Acid Synthase/FASN Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB59271
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB59271 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB59271 Supplier R&D Systems Supplier No. MAB59271
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Fatty Acid Synthase/FASN Monoclonal specifically detects Fatty Acid Synthase/FASN in Human samples. It is validated for Western Blot.

Specifications

Antigen Fatty Acid Synthase/FASN
Applications Western Blot
Classification Monoclonal
Clone 647114
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P49327
Gene Alias EC 2.3.1, EC 2.3.1.85, EC 6.3.3.1, FASN, FASshort chain dehydrogenase/reductase family 27X, member 1, fatty acid synthase, MGC14367, MGC15706, OA-519, SDR27X1
Gene Symbols FASN
Host Species Mouse
Immunogen E. coli-derived recombinant human Fatty Acid Synthase/FASN Ser10-Cys212 Accession # P49327
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2194
Test Specificity Detects Fatty Acid Synthase/FASN in direct ELISAs and Western blots. In direct ELISAs, approximately 25% cross-reactivity with recombinant mouse (rm) Ephrin-A2 is observed and no cross-reactivity with recombinant human (rh) Ephrin-A3, -B3, rmEphrin-A4, -A5, -B1 or -B2 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.