Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Fatty Acid Synthase/FASN Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB59271SP
This item is not returnable.
View return policy
Description
Fatty Acid Synthase/FASN Monoclonal specifically detects Fatty Acid Synthase/FASN in Human samples. It is validated for Western Blot.Specifications
| Fatty Acid Synthase/FASN | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.25 ug/mL | |
| P49327 | |
| FASN | |
| E. coli-derived recombinant human Fatty Acid Synthase/FASN Ser10-Cys212 Accession # P49327 | |
| 25 μg | |
| Primary | |
| Detects Fatty Acid Synthase/FASN in direct ELISAs and Western blots. In direct ELISAs, approximately 25% cross-reactivity with recombinant mouse (rm) Ephrin-A2 is observed and no cross-reactivity with recombinant human (rh) Ephrin-A3, -B3, rmEphrin-A4, -A5, -B1 or -B2 is observed. | |
| Human | |
| Purified |
| Western Blot | |
| 647114 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 2.3.1, EC 2.3.1.85, EC 6.3.3.1, FASN, FASshort chain dehydrogenase/reductase family 27X, member 1, fatty acid synthase, MGC14367, MGC15706, OA-519, SDR27X1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2194 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction