Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human FBXO28 Full-length ORF (NP_055991.1, 1 a.a. - 368 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89944194
Click to view available options
Quantity:
2 μg

Used for AP, Array, ELISA, WB-Re

Members of the F-box protein family, such as FBXO28, are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1 (MIM 601434), cullin (see CUL1; MIM 603134), and F-box proteins, act as protein-ubiquitin ligases. F-box proteins interact with SKP1 through the F box, and they interact with ubiquitination targets through other protein interaction domains (Jin et al., 2004 [PubMed 15520277]).[supplied by OMIM]

Sequence: MAAAAEERMAEEGGGGQGDGGSSLASGSTQRQPPPPAPQHPQPGSQALPAPALAPDQLPQNNTLVALPIVAIENILSFMSYDEISQLRLVCKRMDLVCQRMLNQGFLKVERYHNLCQKQVKAQLPRRESERRNHSLARHADILAAVETRLSLLNMTFMKYVDSNLCCFIPGKVIDEIYRVLRYVNSTRAPQRAHEVLQELRDISSMAMEYFDEKIVPILKRKLPGSDVSGRLMGSPPVPGPSAALTTMQLFSKQNPSRQEVTKLQQQVKTNGAGVTVLRREISELRTKVQEQQKQLQDQDQKLLEQTQIIGEQNARLAELERKLREVMESAVGNSSGSGQNEESPRKRKKATEAIDSLRKSKRLRNRK

Specifications

Accession Number NP_055991.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 23219
Molecular Weight (g/mol) 67.5kDa
Name FBXO28 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 2 μg
Immunogen MAAAAEERMAEEGGGGQGDGGSSLASGSTQRQPPPPAPQHPQPGSQALPAPALAPDQLPQNNTLVALPIVAIENILSFMSYDEISQLRLVCKRMDLVCQRMLNQGFLKVERYHNLCQKQVKAQLPRRESERRNHSLARHADILAAVETRLSLLNMTFMKYVDSNLCCFIPGKVIDEIYRVLRYVNSTRAPQRAHEVLQELRDISSMAMEYFDEKIVPILKRKLPGSDVSGRLMGSPPVPGPSAALTTMQLFSKQNPSRQEVTKLQQQVKTNGAGVTVLRREISELRTKVQEQQKQLQDQDQKLLEQTQIIGEQNARLAELERKLREVMESAVGNSSGSGQNEESPRKRKKATEAIDSLRKSKRLRNRK
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias FLJ10766/Fbx28/KIAA0483
Common Name FBXO28
Gene Symbol FBXO28
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.