Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Fc gamma RII/CD32 Alexa Fluor™ 405-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB1330V100UG
This item is not returnable.
View return policy
Description
Fc gamma RII/CD32 Monoclonal specifically detects Fc gamma RII/CD32 in Human samples. It is validated for Flow Cytometry.Specifications
Fc gamma RII/CD32 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
P31994 | |
FCGR2A | |
Mouse myeloma cell line NS0-derived recombinant human Fc gamma RIIB Ala46-Pro217 Accession # P31994 | |
RUO | |
2213 | |
Human | |
IgG2a |
Flow Cytometry | |
190723 | |
Alexa Fluor 405 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
CD32 antigen, CD32a, CD32b, CD32Fc fragment of IgG, low affinity II, receptor for (CD32), CDw32, Fc fragment of IgG, low affinity IIb, receptor (CD32), Fc fragment of IgG, low affinity IIb, receptor for (CD32), Fc gamma RIIb, FCG2, Fc-gamma RII-b, fc-gamma-RIIb, FCGR2, FCGR2A, FCGR2B, FcgRII, FcgRIIA, FcgRIIB, FCRIIA, fcRII-b, FCRIIB, IGFR2, IgG Fc receptor II-b, low affinity immunoglobulin gamma Fc region receptor II-b | |
Mouse | |
100 μg | |
Primary | |
Detects human Fc gamma RIIA and human Fc gamma RIIB in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant mouse Fc gamma RII. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction