Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Fc gamma RII/CD32 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems BAF1330
This item is not returnable.
View return policy
Description
Fc gamma RII/CD32 Polyclonal specifically detects Fc gamma RII/CD32 in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
| Fc gamma RII/CD32 | |
| Polyclonal | |
| Biotin | |
| P31994 | |
| FCGR2A | |
| Mouse myeloma cell line NS0-derived recombinant human Fc gamma RIIB Ala46-Pro217 Accession # P31994 | |
| 50 μg | |
| Primary | |
| Detects human Fc gamma RII/CD32 in Western blots. In Western blots, approximately 20% cross-reactivity with recombinant mouse CD32 is observed. | |
| Human | |
| IgG |
| Western Blot, Immunocytochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| CD32 antigen, CD32B, CD32Fc fragment of IgG, low affinity II, receptor for (CD32), CDw32, Fc fragment of IgG, low affinity IIb, receptor (CD32), Fc fragment of IgG, low affinity IIb, receptor for (CD32), Fc gamma RIIb, FCG2, Fc-gamma RII-b, fc-gamma-RIIb, FCGR2, FcgRII, FcRII-b, IGFR2, IgG Fc receptor II-b, low affinity immunoglobulin gamma Fc region receptor II-b | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 2213 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction