Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Fc gamma RIIB/C (CD32b/c) Antibody, R&D Systems™
GREENER_CHOICESDP

Catalog No. MAB18751SP Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB18751SP 25 μg
MAB18751100 100 μg
MAB18751500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB18751SP Supplier R&D Systems Supplier No. MAB18751SP
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Fc gamma RIIB/C (CD32b/c) Monoclonal specifically detects Fc gamma RIIB/C (CD32b/c) in Human samples. It is validated for ELISA Capture (Matched Antibody Pair)∣Immunohistochemistry (Paraffin)∣Neutralization.

Specifications

Antigen Fc gamma RIIB/C (CD32b/c)
Applications ELISA Capture (Matched Antibody Pair), Immunohistochemistry (Paraffin), Neutralization
Classification Monoclonal
Clone 190710
Concentration LYOPH
Conjugate Unconjugated
Dilution ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P31994
Gene Symbols FCGR2B
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Fc gamma RIIB/C Ala46-Pro217 Accession # P31994
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2213
Test Specificity Detects human Fc gamma RIIB/C in ELISAs. In a sandwich ELISA shows approximately 40% cross-reactivity with recombinant human (rh) Fc gamma RIIA and no cross-reactivity with rhFc gamma RI, rmFc gamma RI, rmFc gamma RIIB, rhFc gamma RIIIB, rmFc gamma RIII, or rmFcrl3/CD16-2.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.