Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Fc gamma RIII (CD16) Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB2546SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2546SP 25 μg
MAB2546100 100 μg
MAB2546500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB2546SP Supplier R&D Systems Supplier No. MAB2546SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 6 publications

Fc gamma RIII (CD16) Monoclonal specifically detects Fc gamma RIII (CD16) in Human samples. It is validated for Flow Cytometry, CyTOF-ready.

Specifications

Antigen Fc gamma RIII (CD16)
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 245536
Conjugate Unconjugated
Dilution Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. O75015
Gene Alias CD16, CD16A, CD16b, Fc fragment of IgG receptor IIIa, FCG3, FCGR3, FCGR3A, FCGR3B, FCGRIII, FcgRIIIA, FcgRIIIB, FcR-10, FCRIII, FCRIIIA, FCRIIIB, IGFR3, IMD20
Gene Symbols FCGR3B
Host Species Mouse
Immunogen Raised against Fc gamma RIII (CD16).
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2214
Test Specificity Detects human Fc gamma RIIIA/B (CD16) in direct ELISAs. In direct ELISAs, approximately 10% cross-reactivity with recombinant human Fc gamma RIIA and recombinant mouse Fc gamma RIII is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.