Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Fc gamma RIIIA/CD16a Antibody, R&D Systems™
Rabbit Monoclonal Antibody
Specifications
Antigen | Fc gamma RIIIA/CD16a |
---|---|
Clone | 2829B |
Dilution | Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 3-25 ug/mL, Immunocytochemistry 3-25 ug/mL |
Applications | Flow Cytometry, Immunohistochemistry, Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB107512SP
|
R&D Systems
MAB107512SP |
25 μg | N/A | N/A | N/A | ||||
MAB10751210
|
R&D Systems
MAB107512100 |
100 μg | N/A | N/A | N/A | ||||
Description
Fc gamma RIIIA/CD16a Monoclonal antibody specifically detects Fc gamma RIIIA/CD16a in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, ImmunocytochemistrySpecifications
Fc gamma RIIIA/CD16a | |
Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 3-25 ug/mL, Immunocytochemistry 3-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
CD16a, CD16a antigen, CD16FCRIIIA, Fc fragment of IgG, low affinity IIIa, receptor (CD16a), Fc fragment of IgG, low affinity IIIa, receptor for (CD16), Fc gamma receptor III-A, FCG3, Fc-gamma receptor III-2 (CD 16), Fc-gamma receptor IIIb (CD16), Fc-gamma RIII, Fc-gamma RIIIa, Fc-gamma RIII-alpha, FCGR3, FCGR3A, FCGRIII, FcgRIIIA, FCR-10, FcRIII, FcRIIIa, IGFR3FcgammaRIIIA, immunoglobulin G Fc receptor III, low affinity III, receptor for (CD16), low affinity immunoglobulin gamma Fc region receptor III-A, neutrophil-specific antigen NA | |
Mouse myeloma cell line NS0-derived human Fc gamma RIIIA/CD16a, Gly17-Gln208, Accession # P08637 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
2829B | |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Human | |
P08637 | |
2214 | |
IgG | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title