Learn More
Novus Biologicals™ Human Fc gamma RIIIA/CD16a ELISA Kit (Colorimetric)
Shop All R&D Systems Products

Specifications
Specifications
| Assay Range | 7.81 to 500 ng/mL |
| Assay Sensitivity | 4.69 ng/mL |
| Product Type | ELISA Kit (Colorimetric) |
| Sample Type | Cell Culture Supernatant, Other Biological Fluids, Plasma, Serum |
| Specificity | This kit recognizes Human Fc gamma RIIIA/CD16a in samples. No significant cross-reactivity or interference between Human Fc gamma RIIIA/CD16a and analogues was observed. |
| Target Species | Human |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 2214 |
| Gene Symbol | FCGR3A |
| Quantity | 1 Kit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.