Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Fc mu R/FAIM3 Alexa Fluor 405-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB9494V100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB9494V100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB9494V100 Supplier R&D Systems Supplier No. FAB9494V100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

FAIM3 Monoclonal specifically detects FAIM3 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen FAIM3
Applications Flow Cytometry
Classification Monoclonal
Clone 992338
Conjugate Alexa Fluor 405
Dilution Flow Cytometry 0.25-1 μg/106 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Accession No. O60667
Gene Alias FAIM3, Fas apoptotic inhibitory molecule 3, Fc mu receptor, FCMR, immunoglobulin mu Fc receptor, Regulator of Fas-induced apoptosis Toso, Toso, TOSOIgM Fc receptor
Host Species Mouse
Immunogen Human embryonic kidney cell line HEK293-derived recombinant human Fc mu R/FAIM3, Arg18-Gly251, Accession No. O60667
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 9214.0
Test Specificity Detects human Fc mu R/FAIM3 in direct ELISAs. Detects human, mouse, and rat Fc mu R/FAIM3 in Western blots.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Form Supernatant
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.