Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human FCRL1/FcRH1 Alexa Fluor 405-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB2049V100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB2049V100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB2049V100 Supplier R&D Systems Supplier No. FAB2049V100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

FCRL1/FcRH1 Monoclonal specifically detects FCRL1/FcRH1 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen FCRL1/FcRH1
Applications Flow Cytometry
Classification Monoclonal
Clone 282415
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 405
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. Q96LA6
Gene Alias BXMH1, CD307a, CD307a antigen, Fc receptor homolog 1, Fc receptor-like 1, FcRH1, FCRH1DKFZp667O1421, FcRL1, FcR-like protein 1, hIFGP1, IFGP family protein 1, IFGP1, IFGP1Fc receptor-like protein 1, Immune receptor translocation-associated protein 5, IRTA5, IRTA5immunoglobulin superfamily Fc receptor, gp42
Gene Symbols FCRL1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human FCRL1/FcRH1 Ala17-His304 Accession # Q96LA6
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 115350
Test Specificity Detects human FCRL1/FcRH1 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) FCRL2, 3, 4, or 5 is observed.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.