Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human FCRL1/FcRH1 Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $549.50
Specifications
Antigen | FCRL1/FcRH1 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
FCRL1/FcRH1 Polyclonal specifically detects FCRL1/FcRH1 in Human samples. It is validated for Western Blot.Specifications
FCRL1/FcRH1 | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Human | |
BXMH1, CD307a, CD307a antigen, Fc receptor homolog 1, Fc receptor-like 1, FcRH1, FCRH1DKFZp667O1421, FcRL1, FcR-like protein 1, hIFGP1, IFGP family protein 1, IFGP1, IFGP1Fc receptor-like protein 1, Immune receptor translocation-associated protein 5, IRTA5, IRTA5immunoglobulin superfamily Fc receptor, gp42 | |
FCRL1 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
Q96LA6 | |
115350 | |
Mouse myeloma cell line NS0-derived recombinant human FCRL1/FcRH1 Ala17-His304 Accession # Q96LA6 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human FCRL1/FcRH1 in direct ELISAs and Western blots. In Western blots, approximately 10% cross-reactivity with recombinant human (rh) ITRA4 is observed, and less than 5% cross-reactivity with rhIRTA3, rhIRTA2, and rhIRTA1 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title