Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human FCRL3/FcRH3 Alexa Fluor 750-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB3126S100UG
This item is not returnable.
View return policy
Description
FCRL3/FcRH3 Monoclonal specifically detects FCRL3/FcRH3 in Human samples. It is validated for Flow Cytometry.Specifications
FCRL3/FcRH3 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q96P31 | |
FCRL3 | |
Mouse myeloma cell line NS0-derived recombinant human FCRL3/FcRH3 Arg14-Arg569 Accession # Q96P31 | |
RUO | |
115352 | |
Human | |
IgG1 |
Flow Cytometry | |
546828 | |
Alexa Fluor 750 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
CD307c, Fc receptor homolog 3, Fc receptor-like 3, Fc receptor-like protein 3, FCRH3, fcR-like protein 3, hIFGP3, IFGP3, immunoglobulin superfamily receptor translocation associated protein 3, IRTA3, SPAP2 | |
Mouse | |
100 μg | |
Primary | |
Detects human FCRL3/FcRH3 in direct ELISAs. In direct ELISAs, less than 5% cross-reactivity with recombinant human (rh) FCRL2 and rhFCRL5 is observed, and no cross-reactivity with rhFCRL1 or recombinant mouse FCRL3 is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction