Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human FCRL3/FcRH3 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF3126SP
This item is not returnable.
View return policy
Description
FCRL3/FcRH3 Polyclonal specifically detects FCRL3/FcRH3 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
FCRL3/FcRH3 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD307c, Fc receptor homolog 3, Fc receptor-like 3, Fc receptor-like protein 3, FCRH3, fcR-like protein 3, hIFGP3, IFGP3, immunoglobulin superfamily receptor translocation associated protein 3, IRTA3, SPAP2 | |
Goat | |
Affinity Purified | |
RUO | |
115352 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Flow Cytometry, CyTOF | |
LYOPH | |
Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
NP_443171 | |
FCRL3 | |
Mouse myeloma cell line NS0-derived recombinant human FCRL3/FcRH3 Arg14-Arg569 Accession # NP_443171 | |
25 μg | |
Primary | |
Detects human FCRL3/FcRH3 in direct ELISAs. In direct ELISAs, approximately 15% cross-reactivity with recombinant human (rh) IRTA2 and rhIRTA5 is observed. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction