Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human FCRL5/FCRL3 Alexa Fluor 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB20871R100UG
This item is not returnable.
View return policy
Description
FCRL5/FCRL3 Monoclonal specifically detects FCRL5/FCRL3 in Human samples. It is validated for Flow Cytometry.Specifications
| FCRL5/FCRL3 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| AAI01067 | |
| Mouse myeloma cell line NS0-derived recombinant human FCRL5/FCRL3 Gln16-Arg844 Accession # AAI01067 | |
| RUO | |
| Detects human FCRL5/FCRL3 in direct ELISAs. In direct ELISAs, 100% cross-reactivity with recombinant human (rh) FCRL3 is observed and no cross-reactivity with rhFCRL1, 2, or 4 is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
| Flow Cytometry | |
| 307307 | |
| Alexa Fluor 647 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction