Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human FCRL5/FCRL3 Alexa Fluor 750-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FA20871S100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FA20871S100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FA20871S100 Supplier R&D Systems Supplier No. FAB20871S100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

FCRL5/FCRL3 Monoclonal specifically detects FCRL5/FCRL3 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen FCRL5/FCRL3
Applications Flow Cytometry
Classification Monoclonal
Clone 307307
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 750
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. AAI01067
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human FCRL5/FCRL3 Gln16-Arg844 Accession # AAI01067
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Detects human FCRL5/FCRL3 in direct ELISAs. In direct ELISAs, 100% cross-reactivity with recombinant human (rh) FCRL3 is observed and no cross-reactivity with rhFCRL1, 2, or 4 is observed.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2b
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.