Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human FCRL5/FCRL3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB20871SP
This item is not returnable.
View return policy
Description
FCRL5/FCRL3 Monoclonal specifically detects FCRL5/FCRL3 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
FCRL5/FCRL3 | |
Monoclonal | |
LYOPH | |
Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
AAI01067 | |
Mouse myeloma cell line NS0-derived recombinant human FCRL5/FCRL3 Gln16-Arg844 Accession # AAI01067 | |
25 μg | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Flow Cytometry, CyTOF | |
307307 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Detects human FCRL5/FCRL3 in direct ELISAs. In direct ELISAs, 100% cross-reactivity with recombinant human (rh) FCRL3 is observed and no cross-reactivity with rhFCRL1, 2, or 4 is observed. | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction