Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human FCRL6/FcRH6 Alexa Fluor 405-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB6745V100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB6745V100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB6745V100 Supplier R&D Systems Supplier No. FAB6745V100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

FCRL6/FcRH6 Monoclonal specifically detects FCRL6/FcRH6 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen FCRL6/FcRH6
Applications Flow Cytometry
Classification Monoclonal
Clone 672505
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 405
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. Q6DN72
Gene Alias Fc receptor-like 6, FcRH6, FLJ16056, IFGP6
Gene Symbols FCRL6
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human FCRL6/FcRH6 Lys16-Leu312 Accession # Q6DN72
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 343413
Test Specificity Detects human FCRL6/FcRH6 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) FCRL1, rhFCRL2, rhFCRL3, rhFCRL4, rhFCRL5, rhFcRN, or rhFCRLB/FCRY is observed.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.