Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human FCRL6/FcRH6 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB6745
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB6745 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB6745 Supplier R&D Systems Supplier No. MAB6745
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

FCRL6/FcRH6 Monoclonal specifically detects FCRL6/FcRH6 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.

Specifications

Antigen FCRL6/FcRH6
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 672505
Concentration LYOPH
Conjugate Unconjugated
Dilution Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q6DN72
Gene Alias Fc receptor-like 6, FcRH6, FLJ16056, IFGP6
Gene Symbols FCRL6
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human FCRL6/FcRH6 Lys16-Leu312 Accession # Q6DN72
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 343413
Test Specificity Detects human FCRL6/FcRH6 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) FCRL1, rhFCRL2, rhFCRL3, rhFCRL4, rhFCRL5, rhFcRN, or rhFCRLB/FCRY is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.