Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human FCRLB/FCRY Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF4705
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF4705 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF4705 Supplier R&D Systems Supplier No. AF4705
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

FCRLB/FCRY Polyclonal specifically detects FCRLB/FCRY in Human samples. It is validated for Western Blot.

Specifications

Antigen FCRLB/FCRY
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Gene Accession No. Q6BAA4
Gene Alias Fc receptor homolog expressed in B cells protein 2, Fc receptor like 2, Fc receptor-like and mucin-like 2, Fc receptor-like and mucin-like protein 2, Fc receptor-like B, Fc receptor-like protein 2, Fc receptor-related protein Y, FcRL2, FCRL2FCRLM2, FCRLM2, FCRLY, FCRY, FLJ31052, FREB2, FREB-2, FREB-2FcRY, RP11-474I16.6
Gene Symbols FCRLB
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant human FCRLB/FCRY Ala18-Ser426 Accession # Q6BAA4
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 127943
Test Specificity Detects human FCRLB/FCRY in direct ELISAs and Western blots. In direct ELISAs, approximately 30% cross-reactivity with recombinant mouse (rm) FCRLB/FCRY is observed, and less than 1% cross-reactivity with recombinant human FCRN and rmFCRN is observed.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.