Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Human Femur Bone Model
SCI_ED

Catalog No. p-7201182
Click to view available options
Includes:
Left Side
Right Side

Life-size human femur bone provides an incredible amount of realism and detail for examining the human skeleton

  • Life-size, anatomically correct replica
  • Realistic details, textures, and bony landmarks
  • Made of durable and sturdy PVC plastic
  • Ideal for both student and patient educational demonstrations
  • Washable

Specifications

Product Type Anatomical Model, Human
For Use With (Application) Examining The Human Skeleton
Includes Left Side
Length (English) 16 in.
Material PVC
Width (English) 3.5 in.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.