Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Ferroportin/SLC40A1 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$214.00 - $514.50
Specifications
Antigen | Ferroportin/SLC40A1 |
---|---|
Clone | 1308C |
Dilution | Flow Cytometry 0.25 μg/mL, Immunohistochemistry 1-25 μg/mL, CyTOF-ready |
Applications | Flow Cytometry, Immunohistochemistry, CyTOF |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB9924SP
![]() |
R&D Systems
MAB9924SP |
25 μg |
Each for $214.00
|
|
|||||
MAB9924100
![]() |
R&D Systems
MAB9924100 |
100 μg |
Each for $514.50
|
|
|||||
Description
Ferroportin/SLC40A1 Monoclonal specifically detects Ferroportin/SLC40A1 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF-ready.Specifications
Ferroportin/SLC40A1 | |
Flow Cytometry 0.25 μg/mL, Immunohistochemistry 1-25 μg/mL, CyTOF-ready | |
Monoclonal | |
Supernatant | |
Human | |
Q9NP59 | |
30061 | |
IgG | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
1308C | |
Flow Cytometry, Immunohistochemistry, CyTOF | |
Unconjugated | |
Rabbit | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
Ferroportin-1, FPN1, FPN1IREG1ferroportin 1, HFE4, HFE4ferroportin-1, IREG1, iron regulated gene 1, Iron-regulated transporter 1, member 3, MST079, MSTP079, MTP1, putative ferroportin 1 variant IIIB, SLC11A3, SLC11A3iron regulated gene 1, SLC40A1, solute carrier family 11 (proton-coupled divalent metal ion transporters), solute carrier family 11 (proton-coupled divalent metal ion transporters), member 3, solute carrier family 40 (iron-regulated transporter), member 1, solute carrier family 40 member 1 | |
Synthetic peptide containing human Ferroportin/SLC40A1 Accession # Q9NP59 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Ferroportin/SLC40A1 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title