Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Fetuin B Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB17251SP
This item is not returnable.
View return policy
Description
Fetuin B Monoclonal specifically detects Fetuin B in Human samples. It is validated for Western Blot, Simple Western, ELISA Capture (Matched Antibody Pair).Specifications
Fetuin B | |
Monoclonal | |
LYOPH | |
Western Blot 5 ug/mL, Simple Western 50 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
Q9UGM5 | |
FETUB | |
Mouse myeloma cell line NS0-derived recombinant human Fetuin B Met19-Pro382 Accession # Q9UGM5 | |
25 μg | |
Primary | |
Detects human Fetuin B in ELISAs. In sandwich immunoassays, no cross-reactivity or interference with recombinant human (rh) Fetuin A, rhCystatin A, B, C, rhMMP-2, 9, rhBMP-2, 4, 6, rhTGF-beta 1, or rhTGF-beta 2 is observed. | |
Human | |
Purified |
Western Blot, ELISA | |
212607 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
FETUB, fetuin-B, Fetuin-like protein IRL685,16G2, Gugu, IRL685 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
26998 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction