Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Fetuin B Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAM17251
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAM17251 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAM17251 Supplier R&D Systems Supplier No. BAM17251
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Fetuin B Monoclonal specifically detects Fetuin B in Human samples. It is validated for ELISA Detection (Matched Antibody Pair).

Specifications

Antigen Fetuin B
Applications ELISA
Classification Monoclonal
Clone 212633
Concentration LYOPH
Conjugate Biotin
Dilution ELISA Detection (Matched Antibody Pair) 0.5-2.0 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein. with No Preservative
Gene Accession No. NP_055190
Gene Alias FETUB, fetuin-B, Fetuin-like protein IRL685,16G2, Gugu, IRL685
Gene Symbols FETUB
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Fetuin B Met19-Pro382 Accession # NP_055190
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 250 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26998
Test Specificity Detects human Fetuin B in ELISAs. In sandwich immunoassays, no cross-reactivity or interference with recombinant human (rh) Fetuin A, rhCystatin A, B, C, rhMMP-2, 9, rhBMP-2, 4, 6, rhTGF-beta 1, or rhTGF-beta 2 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.