Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Fetuin B Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems BAF1725
This item is not returnable.
View return policy
Description
Fetuin B Polyclonal specifically detects Fetuin B in Human samples. It is validated for Western Blot.Specifications
Fetuin B | |
Polyclonal | |
Biotin | |
Q9UGM5 | |
FETUB | |
Mouse myeloma cell line NS0-derived recombinant human Fetuin B Met19-Pro382 Accession # Q9UGM5 | |
50 μg | |
Primary | |
Detects human Fetuin B in Western blots. In Western blots, 2% cross-reactivity with recombinant human (rh) Fetuin A, rhCystatin A, rhCystatin B, rhCystatin C, rhCystatin D, rhCystatin E/M, rhCystatin S, rhCystatin SA, and rhCystatin SN. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
FETUB, fetuin-B, Fetuin-like protein IRL685,16G2, Gugu, IRL685 | |
Goat | |
Affinity Purified | |
RUO | |
26998 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction