Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human FFAR2/GPR43 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $502.50
Specifications
| Antigen | FFAR2/GPR43 |
|---|---|
| Clone | 1000624 |
| Dilution | Flow Cytometry 0.25 μg/106 cells, CyTOF-ready |
| Applications | Flow Cytometry, CyTOF |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB10082SP
![]() |
R&D Systems
MAB10082SP |
25 μg |
Each for $200.50
|
|
|||||
MAB10082100
![]() |
R&D Systems
MAB10082100 |
100 μg |
Each for $502.50
|
|
|||||
Description
FFAR2/GPR43 Monoclonal specifically detects FFAR2/GPR43 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
| FFAR2/GPR43 | |
| Flow Cytometry 0.25 μg/106 cells, CyTOF-ready | |
| Monoclonal | |
| Supernatant | |
| Human | |
| O15552 | |
| 2867 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
| 1000624 | |
| Flow Cytometry, CyTOF | |
| Unconjugated | |
| Mouse | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| free fatty acid activated receptor 2, free fatty acid receptor 2, G protein-coupled receptor 43, GPR43, GPR43FFA2R, G-protein coupled receptor 43 | |
| NS0 Mouse myeloma cell line transfected with human FFAR2/GPR43, Met1-Glu330, Accession # O15552 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Stains human FFAR2/GPR43 transfectants but not irrelevant transfectants in flow cytometry. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title